Organic reference standard, 2,3',4,4'-Tetrachlorobiphenyl (PCB No. 66) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-66
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:32598-10-0
- Description:2,3',4,4'-Tetrachlorobiphenyl (PCB No. 66) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₆Cl₄
- Description:2,3',4,4'-Tetrachlorobiphenyl (PCB No. 66)