Order Entry
Spain
ContactUsLinkComponent
 
Organic reference standard, 2,2',4,4',6,6'-Hexachlorobiphenyl (PCB No. 155) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-155-CS
 :  
Organic reference standard, 2,2',4,4',6,6'-Hexachlorobiphenyl (PCB No. 155) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-155-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    33979-03-2
  • Description:
    2,2',4,4',6,6'-Hexachlorobiphenyl (PCB No. 155) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',4,4',6,6'-Hexachlorobiphenyl (PCB No. 155)