Organic reference standard, 3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-77
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:32598-13-3
- Description:3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₆Cl₄
- Description:3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77)