Organic reference standard, 2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-149-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:38380-04-0
- Description:2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 100 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149)