Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, 2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-153-CS
 :  
Organic reference standard, 2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-153-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    35065-27-1
  • Description:
    2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)