Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, 2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-191
 :  
Organic reference standard, 2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-191
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    74472-50-7
  • Description:
    2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,3,3',4,4',5',6-Heptachlorobiphenyl (PCB No. 191)