Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-196-CS
 :  
Organic reference standard, 2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-196-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    42740-50-1
  • Description:
    2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,2',3,3',4,4',5,6'-Octachlorobiphenyl (PCB No. 196)