Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-194
 :  
Organic reference standard, 2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-194
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    35694-08-7
  • Description:
    2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194) 35 µg/ml in isooctane
  • Environmentally Preferable:
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194)