Pesticide reference standard, 1,4-Dichlorobenzene 5000 µg/ml in methanol, Supelco®
Catalog # SUPL40025
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- CAS No.:106-46-7
- Description:1,4-Dichlorobenzene 5,000 µg/ml in methanol
- Formula:C₆H₄Cl₂
- Description:1,4-Dichlorobenzene