Pesticide reference standard, 1,3-Dichlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12371000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:541-73-1
- Description:1,3-Dichlorobenzene 100 µg/ml in methanol
- Formula:C₆H₄Cl₂
- Description:1,3-Dichlorobenzene