Order Entry
Spain
ContactUsLinkComponent
Pesticide reference standard, 1,2-Dichlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 1,2-Dichlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12370000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1,2-Dichlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12370000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    95-50-1
  • Description:
    1,2-Dichlorobenzene 100 µg/ml in methanol
  • Formula:
    C₆H₄Cl₂
  • Description:
    1,2-Dichlorobenzene

Specifications

About this item