Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Anthracene 100 µg/ml in acetonitrile
Organic reference standard, Anthracene 100 µg/ml in acetonitrile
Catalog # PROONE5011
CAS Number:  
Organic reference standard, Anthracene 100 µg/ml in acetonitrile
Catalog # PROONE5011
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • CAS No.:
    120-12-7
  • Description:
    Anthracene 100 µg/ml in acetonitrile
  • Formula:
    C₁₄H₁₀ 
  • Description:
    Anthracene

Specifications

About this item