Order Entry
Spain
ContactUsLinkComponent
Magnesium perchlorate hydrate for elementary analysis, Supelco®
 
 
Magnesium perchlorate hydrate for elementary analysis, Supelco®
  1.05875.0500
 :  Merck
 :  
 
 
Magnesium perchlorate hydrate for elementary analysis, Supelco®
  1.05875.0500
 :  Merck
 :  

 

  • Pack type:
    Metal can
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Solubility:
    500 g/l (25 °C)
  • Melting Point:
    251 °C decomposes
  • Bulk Density:
    600 - 800 kg/m3
  • Density:
    2.60 g/cm3 (25 °C) (anhydrous substance)
  • pH:
    8.2 (50 g/l, H2O, 25 °C)

 

 

 :
 :241.22 g/mol
 :146 °C
 :2.6 g/cm³ (25 °C) (anhydr.)
 :5.1,II
 :M-Clarity™ quality level = MQ100
 :233-108-3
 :1475
 :GHS03
 :GHS07
 :Ambient
 :64010-42-0
 :Cl₂MgO₈·H₂O