

Magnesium perchlorate hydrate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:64010-42-0
:
Formula:Cl₂MgO₈·H₂O
:241.22 g/mol
Melting Pt:146 °C
Density:2.6 g/cm³ (25 °C) (anhydr.)
Storage Temperature:Ambient
EINECS:233-108-3
UN:1475
ADR:5.1,II
:M-Clarity™ quality level = MQ300
:GHS03
:GHS07
Specification Test Results
- Solubility:500 g/l (25 °C)
- Melting Point:251 °C decomposes
- Bulk Density:600 - 800 kg/m3
- Density:2.60 g/cm3 (25 °C) (anhydrous substance)
- pH:8.2 (50 g/l, H2O, 25 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Frequently Bought Together