


Sodium perchlorate monohydrate 98% ACS, Fluka™
310514-500G
:
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
- Assay:98.0 - 102.0 %
- Assay of NaClO4:85.0 - 90.0 %
- pH (5 %, 25 °C):6.0 - 8.0
- Insoluble matter:Max. 0.005 %
- Calcium (Ca):Max. 200 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 500 ppm
- Heavy metals (as Pb):Max. 5 ppm
- Sulfate (SO₄):Max. 20 ppm
- Chloride (Cl):Max. 30 ppm
:
:140.46 g/mol
:12,08798
:Decomposes on heating
: Dec. (on heating)
:469 °C (anhydr.)
:2.02 g/cm³ (20 °C)
:Ambient
:231-511-9
:1502
:5.1,II
:GHS03
:GHS08
:GHS07
:7791-07-3
:NaClO₄·H₂O
:MFCD00149164