Perclorato de litio trihidrato, grado reactivo
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 13453-78-6 Molecular Formula: ClLiO4.3H2O Formula Weight: 160.45 (106.40anhy) Physical Form: Crystalline Density: 1.84 Appearance: White Odour: Odourless Merck Reference: 14,5539 UN No.: 1481 MDL No.: MFCD00149765
Synonyms: Litio perclorato trihidrato
- CAS Number:13453-78-6
- Formula:ClLiO₄·3H₂O
- MDL Number:MFCD00149765
- Molecular Weight:160.44 g/mol
- Merck Index:14,5539
- Melting Pt:91,85 °C
- Density:1,84
- Storage Temperature:Ambiente
- EINECS:232-237-2
- UN:1481
- ADR:5.1,II
- :Density: 1.841 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
CAS No.: 13453-78-6 Molecular Formula: ClLiO4.3H2O Formula Weight: 160.45 (106.40anhy) Physical Form: Crystalline Density: 1.84 Appearance: White Odour: Odourless Merck Reference: 14,5539 UN No.: 1481 MDL No.: MFCD00149765
Synonyms: Litio perclorato trihidrato
- CAS Number:13453-78-6
- Formula:ClLiO₄·3H₂O
- MDL Number:MFCD00149765
- Molecular Weight:160.44 g/mol
- Merck Index:14,5539
- Melting Pt:91,85 °C
- Density:1,84
- Storage Temperature:Ambiente
- EINECS:232-237-2
- UN:1481
- ADR:5.1,II
- :Density: 1.841 g/cm³ (20 °C)



