Order Entry
Spain
ContactUsLinkComponent
Zinc acetate dihydrate 99.0-101.0% (by complexometric analysis), EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
Corrosive
Irritant
Environment
Zinc acetate dihydrate 99.0-101.0% (by complexometric analysis), EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
Certificates
Corrosive
Irritant
Environment
Zinc acetate dihydrate 99.0-101.0% (by complexometric analysis), EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
Certificates

Specification Test Results

  • Assay (Complexometric):
    99.0 - 101.0%
  • Assay Acetic Acid (Potentiometric):
    ≤0.5%
  • Identity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Insoluble Matter:
    ≤0.005%
  • pH-value (5%; Water):
    6.0 - 7.0
  • Chloride (Cl):
    ≤0.0050%
  • Sulfate (SO₄):
    ≤0.0100%
  • Al (Aluminium):
    ≤0.0005%
  • As (Arsenic):
    ≤0.0002%
  • Cd (Cadmium) (*):
    ≤0.0002%
  • Cu (Copper):
    ≤0.0050%
  • Fe (Iron):
    ≤0.0050%
  • Pb (Lead):
    ≤0.0010%
  • Alkali- and Alkaline Earth Metals:
    ≤0.2%
  • Other Residual Solvents (ICH Q3C):
    Excluded by Manufacturing Process
  • Reducing Substances:
    Passes Test

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together