Order Entry
Spain
ContactUsLinkComponent
L(+)-Histidine monohydrochloride monohydrate
L(+)-Histidine monohydrochloride monohydrate
Catalog # A3733.0100
Supplier:  PanReac AppliChem
CAS Number:  
undefined
L(+)-Histidine monohydrochloride monohydrate
Catalog # A3733.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :209.63 g/mol
Merck Index:13,04741
Melting Pt:259 °C
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-438-9
CAS Number:5934-29-2
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027