
Potassium hydroxide 40% in aqueous solution
Catalog # PRLSMC311938.5000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553