0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Spain
ContactUsLinkComponent
Hydrofluoric acid
Hydrofluoric acid 50%
Hydrofluoric acid 50%
Hydrofluoric acid 50%
HONE17602-5L
:
Honeywell Chemicals
:
Hydrofluoric acid 50%
HONE17602-5L
:
Honeywell Chemicals
:
Pk:
5 L
Seq:
5190
:
:
20.01 g/mol
:
Ambient
:
231-634-8
:
1790
:
8,II
:
GHS06
:
GHS05
:
7664-39-3
:
HF
:
MFCD00011346