Order Entry
Spain
ContactUsLinkComponent
Magnesium chloride 0.1 N in aqueous solution
Magnesium chloride 0.1 N in aqueous solution
Catalog # PRLSMC309807.6010
Supplier:  PROLABO CMP
CAS Number:  
Magnesium chloride 0.1 N in aqueous solution
Catalog # PRLSMC309807.6010
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :95.21 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106