Order Entry
Spain
ContactUsLinkComponent
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
Corrosive
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
Catalog # PRLSMC309806.6010
Supplier:  PROLABO CMP
CAS Number:  
Corrosive
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
Catalog # PRLSMC309806.6010
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553