Order Entry
Spain
ContactUsLinkComponent
Conductivity Standard 147 µS/cm (25 °C; 0,001 mol/L; KCl) for Conductivity Measurement
  PRLS87180.260
 :  
Conductivity Standard 147 µS/cm (25 °C; 0,001 mol/L; KCl) for Conductivity Measurement
  PRLS87180.260
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 147 µS/cm (0,001 mol/l; KCl)
  • Environmentally Preferable: