Order Entry
Spain
ContactUsLinkComponent
Acetic acid ≥80%, TECHNICAL
Acetic acid ≥80%, TECHNICAL
Catalog # 20119.368
Supplier:  VWR Chemicals
CAS Number:  
undefined
Acetic acid ≥80%, TECHNICAL
Catalog # 20119.368
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay:
    78 - 82 %
  • Density (20/4):
    1.068 - 1.070

Specification Test Results

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,II
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152