Acetic acid 80% ≥80%
Supplier: PROLABO CMP
Jump to:
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Storage Temperature:Ambient
- EINECS:200-580-7
- UN:2789
- ADR:8,II
3 Options Available Below
Product Details & Documents
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Storage Temperature:Ambient
- EINECS:200-580-7
- UN:2789
- ADR:8,II
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



