Order Entry
Spain
ContactUsLinkComponent
 
Irritant
Health
Chloroform ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650498-1L
CAS Number:  
undefined
Irritant
Health
Chloroform ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650498-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • 2-methyl-2-buten (GC):
    20 - 200 ppm
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 275 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.30
  • Absorbance at 245 nm:
    Max. 1.00
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • Contains:
    amylenes as stabilizer
  • Refractive Index:
    n20/D 1.445 (lit.)

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61 °C (1.013 hPa)
Melting Pt:-61.3 °C
Melting Pt:–61.3 °C
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with amylene
REACH:05-2114614906-45
 :GHS07
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826