

Dichloromethane ≥99.7% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:14,6063
Boiling Pt:39…40 °C
Melting Pt:–95 °C
Density:1.325
UN:1593
ADR:6.1,III
:Stabilised with amylene
EINECS:200-838-9
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together