Polyvinylpyrrolidone (K15), BioChemica®
Catalog # A2258.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (from N):12.0 - 12.8 %
- pH (5 %; H2O):3.0 - 5.0
- Heavy metals (as Pb):max. 0.001 %
- Water (K.F.):max. 5 %
- Solubility (10 %; H2O):clear, colorless to yellowish
Specification Test Results
About this item
:
Storage Temperature:Ambient
CAS Number:9003-39-8
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626