Tannic acid 95%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Gallotannin, Tannin
- CAS Number:1401-55-4
- Formula:C₇₆H₅₂O₄₆
- MDL Number:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Melting Pt:218 °C (dec.)
- Density:Min, 1 g/cm³ (20 °C)
- Flash Pt:198 °C
- Storage Temperature:Ambient
- EINECS:215-753-2
4 Options Available Below
Product Details & Documents
Synonyms: Gallotannin, Tannin
- CAS Number:1401-55-4
- Formula:C₇₆H₅₂O₄₆
- MDL Number:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Melting Pt:218 °C (dec.)
- Density:Min, 1 g/cm³ (20 °C)
- Flash Pt:198 °C
- Storage Temperature:Ambient
- EINECS:215-753-2
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Assay:≥94 % (on dry substance)
- Loss on drying:≤12.0 %
- Sulfated ash:≤1.0 %



