Polyethylene glycol 3,000, Supelco®
Catalog # 81291-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:3000 g/mol
Melting Pt:55…58 °C
Density:1.21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH