Polyethylene glycol 20,000 for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
:20000 g/mol
Melting Pt:58…63 °C
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:500-038-2
Specification Test Results
- Solubility:500 g/l (20 °C)
- Melting Point:58 - 63 °C
- Molar Mass:20000 g/mol
- Bulk Density:400 - 500 kg/m3
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:260 °C
- Density:1.2 g/cm3 (20 °C)
- pH:4 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:320 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.18897.1000_TDS_EN ProductPage_818897
Product specification
msds_818897
SDS
Frequently Bought Together