Order Entry
Spain
ContactUsLinkComponent
Gum arabic, spray-dried, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Gum arabic, spray-dried, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Catalog # 1.04228.9015
CAS Number:  
Gum arabic, spray-dried, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Catalog # 1.04228.9015
CAS Number:  

Specifications

  • Pk:
    15 kg
  • Seq:
    4211

Specifications

About this item

 :
Density:1.35 g/cm³ (20 °C)
Flash Pt:>250 °C
EINECS:232-519-5
 :GHS07
Storage Temperature:Ambient
CAS Number:9000-01-5
MDL Number:MFCD00081264