Gum arabic (total ash max 4%)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9000-01-5
:
MDL Number:MFCD00081264
Merck Index:14,14
Density:1.4
Flash Pt:>250 °C
EINECS:232-519-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together