Order Entry
Spain
Orders LinkContactUsLinkComponent
Agar

Agar

Marketsource
Supplier:   DIFCO BD
Agar
Supplier:  DIFCO BD
Marketsource
Certificates
Agar

2 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 82,45 €
    82,45 €
    1 * 500 g
       82,45 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    2 kg
    Log in for availability
    Log in for availability
    1 * 2 kg
    1 * 2 kg - 588,21 €
    588,21 €
    1 * 2 kg
       588,21 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.