Order Entry
Spain
Orders LinkContactUsLinkComponent
 

D-(+)-Galactose

Plastic bottle for solids, 100 g
Catalog # USBIG1030-100
Supplier:   US Biological
D-(+)-Galactose
Catalog # USBIG1030-100
Supplier:  US Biological
CAS Number:  59-23-4
D-(+)-Galactose
D-(+)-Galactose

Plastic bottle for solids, 100 g

CAS Number:  59-23-4
Price
132,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:163…169 °C
  • Density:1.5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.