Order Entry
Spain
Orders LinkContactUsLinkComponent
D-(+)-Maltose monohydrate

D-(+)-Maltose monohydrate

Glass bottle for solids, 500 g
Catalog # DIFC216830
Marketsource
Supplier:   DIFCO BD
D-(+)-Maltose monohydrate
Catalog # DIFC216830
Supplier:  DIFCO BD
Marketsource
CAS Number:  6363-53-7
D-(+)-Maltose monohydrate
D-(+)-Maltose monohydrate

Glass bottle for solids, 500 g

CAS Number:  6363-53-7
Price
159,58 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
  • CAS Number:6363-53-7
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • MDL Number:MFCD00149343
  • Molecular Weight:360.32 g/mol
  • Merck Index:13,05736
  • Melting Pt:160…165 °C
  • Storage Temperature:Ambient
  • EINECS:200-716-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications