Order Entry
Spain
ContactUsLinkComponent
 
 
 
 
Benzalkonium chloride alkyl distribution from C8H17 to C16H33
 
undefined
 
 
 
Benzalkonium chloride alkyl distribution from C8H17 to C16H33
 

 

  • Appearance:
    Colorless to white to yellow Solid or thick gel (May contain yellowish-white fragments)
  • Infrared spectrum:
    Conforms
  • Assay:
    95.0 to 104.0 % (on anhydrous basis)
  • Water:
    ≤10.0 % (K.F.)
  • Sulfated ash:
    ≤0.5 %
  • Total amines:
    ≤3.6 %

 

Frequently Bought Together