Order Entry
Spain
ContactUsLinkComponent
Phenanthrene-[D10]
Phenanthrene-[D10]
Catalog # EHERC20920100
Supplier:  EHRENSTORFER
CAS Number:  
Phenanthrene-[D10]
Catalog # EHERC20920100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :188.15 g/mol
CAS Number:1517-22-2
Formula:C₁₄D₁₀