Ethyl oleate mixture of esters stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-62-6
:85049-36-1
Formula:C₂₀H₃₈O₂
MDL Number:MFCD00009579
:310.52 g/mol
Density:0.87 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-32 °C
- Boiling Point:168 °C (1.3 hPa)
- Flash Point:110 °C
- Density:0.87 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together