VWR®, Pneumatic Handheld Crimping Tools
548-0304
- Description:Basic pneumatic crimping tool
- Seq:21
- Environmentally Preferable:
- Pk:1
Crimping and decapping tool operated by compressed air (min. 6,2 bar).
- Easy handling: Just by pushing the button the vial is crimped or decapped
- Adjustable and constant crimping pressure
- Interchangeable crimping or decapping heads
- CE mark of conformity
Space-saving installation with balancer (as an option - see ordering table) above the working bench to compensate weight of the crimper. Requires air supply connector with ¼" thread (female). Stand with foot switch is also available as an accessory.