Order Entry
Spain
ContactUsLinkComponent
Iridium ≥99.8% (metals basis), slug, Length: 12.0 mm (0.48 in), Ø 6.0 mm (0.24 in)
Iridium ≥99.8% (metals basis), slug, Length: 12.0 mm (0.48 in), Ø 6.0 mm (0.24 in)
Catalog # 45121.KG
CAS Number:  
Iridium ≥99.8% (metals basis), slug, Length: 12.0 mm (0.48 in), Ø 6.0 mm (0.24 in)
Catalog # 45121.KG
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7439-88-5
 :
Formula:Ir
MDL Number:MFCD00011062
 :192.22 g/mol
Boiling Pt:5300 °C (1013 hPa)
Melting Pt:2450 °C
Storage Temperature:Ambient
EINECS:231-095-9