Molybdenum ≥99.95% (metals basis), plate, Thickness: 4.76 mm (0.188 in)
Catalog # 42143.GH
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
:95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2
CAS Number:7439-98-7
Formula:Mo
MDL Number:MFCD00003465