Molybdenum ≥99.7% (metals basis), pellets
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-98-7
:
Formula:Mo
MDL Number:MFCD00003465
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
:95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2
Related Information
Frequently Bought Together