
Titanium ≥99.99% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-32-6
:
Formula:Ti
MDL Number:MFCD00011264
:47.88 g/mol
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4.51 g/cm³ (20 °C)
EINECS:231-142-3
:GHS02
Storage Temperature:Ambient
Frequently Bought Together