Yttrium ≥99% (REO, rare earth oxide basis), ingot/button, Ø 50.8 mm (2.0 in), Thickness: 11 mm (0.43 in)
Catalog # 45114.KF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:7440-65-5
:
Formula:Y
MDL Number:MFCD00011468
:88.91 g/mol
Merck Index:12,10240
Boiling Pt:2927 °C (1013 hPa)
Melting Pt:1525 °C
Density:4.472 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-174-8