Tin ≥98.8% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
44154.RG
:
- Pk:1
- Seq:7800
- Environmentally Preferable:
- Pk Info:30×150 cm
:7440-31-5
:
:Sn
:MFCD00133862
:118.71 g/mol
:2270
:232
:7.31 g/cm³ (25 °C)
:Ambient
:231-141-8
:12,09587