Standards and test mixes for liquid chromatography
Supplier: Restek
Certificates
About this item
Linear detector responses to concentration variations are an important part of operation qualification (OQ) for LC instruments. Kit of five aqueous solutions of caffeine can be used to generate simple plots of UV response versus concentration.
- Simple check of a detector’s ability to produce a linear response to varied concentrations
- Suitable for clear indication of linear range
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Frequently Bought Together