Order Entry
Spain
ContactUsLinkComponent
R18 peptide
R18 peptide
Catalog # ENZOBMLP2140001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
R18 peptide
Catalog # ENZOBMLP2140001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    R18 Peptide
  • Purity:
    95%

Specifications

About this item

14-3-3 inhibitor

The peptide sequence, PHCVPRDLSWLDLEANMCLP, named R18, was identified as an unphosphorylated peptide that binds to 14-3-3 proteins with high affinity (Kd=80 nM). It has been shown to bind to the same amphipathic groove on the surface of 14-3-3 as phosphorylated peptides. R18 can inhibit the formation of the 14-3-3-Raf-1 complex, reduce Raf-1 dependent transcriptional activity in cells and block the activation of ExoS by 14-3-3. It binds to all the 14-3-3 isoforms with equal affinities. This peptide is useful for competitive inhibition experiments. A biotinylated form is available (BML-P225), allowing detection using streptavidin.