Order Entry
Spain
ContactUsLinkComponent
PKI
PKI
Catalog # ENZOBMLP2030500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PKI
Catalog # ENZOBMLP2030500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Seq:
    02
  • Protein/peptide name:
    PKI
  • Purity:
    >95% (HPLC)

Specifications

About this item

PKA inhibitor

Sequence is from the heat-stable skeletal muscle inhibitor protein of PKA. Peptide binds to the catalytic subunit of PKA and displaces the regulatory subunit. Mimics protein substrate by binding to the catalytic site via the Arg-cluster basic residues. Ki =2,3 nM for PKA.