Order Entry
Spain
ContactUsLinkComponent
 
Protease inhibitor cocktail
Catalog # ENZOBMLKI1030001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Protease inhibitor cocktail
Catalog # ENZOBMLKI1030001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Source:
  • Species:
  • Description:
    Protease inhibitor cocktail
  • Seq:
    02
  • Environmentally Preferable:

Specifications

About this item